Learn More
MAPK2 (ERK2) Mouse anti-Human, Rat, Mouse, Clone: 1B3B9, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | MAPK2 (ERK2) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1B3B9 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. |
| Host Species | Mouse |
| Immunogen | HPLC purified recombinant mouse p42MAP kinase (ERK2). |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.