Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK4 Chicken anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207711
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7711 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7711 Supplier Fabgennix Inc Supplier No. MAPK4101Y
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

MAPK4 Polyclonal antibody specifically detects MAPK4 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen MAPK4
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene MAPK4
Gene Accession No. P31152, Q63454, Q6P5G0
Gene Alias A330097D03Rik; Erk3; Erk3-related; ERK4; ERK-4; Extracellular signal-regulated kinase 4; MAP kinase 4; MAP kinase isoform p63; MAPK 4; MAPK4; mitogen activated protein kinase 4; mitogen-activated protein kinase 4; MNK2; p63Mapk; p63-MAPK; Prkm4
Host Species Chicken
Immunogen Synthetic peptide taken within amino acid region 500-550 on human Mitogen-activated protein kinase 4.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225724, 54268, 5596
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.