Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAPK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112603 Supplier Invitrogen™ Supplier No. PA5112603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAPK4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPK4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAPK4
Gene Accession No. P31152, Q6P5G0
Gene Alias A330097D03Rik; Erk3; Erk3-related; ERK4; ERK-4; Extracellular signal-regulated kinase 4; MAP kinase 4; MAP kinase isoform p63; MAPK 4; MAPK4; mitogen activated protein kinase 4; mitogen-activated protein kinase 4; MNK2; p63Mapk; p63-MAPK; Prkm4
Gene Symbols MAPK4
Host Species Rabbit
Immunogen Recombinant Human Mitogen-activated protein kinase 4 protein (348-587AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225724, 5596
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.