Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK6 Chicken anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207714
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7714 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7714 Supplier Fabgennix Inc Supplier No. MAPK6101Y
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

MAPK6 Polyclonal antibody specifically detects MAPK6 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen MAPK6
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene MAPK6
Gene Accession No. P27704, Q16659, Q61532
Gene Alias 2610021I23Rik; D130053K17Rik; DKFZp686F03189; erk3; ERK-3; Erk-3 related; Extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; HsT17250; LOC538094 protein; MAP kinase 6; MAP kinase isoform p97; MAPK 6; Mapk4; Mapk6; Mapk63; mitogen activated protein kinase 4; mitogen activated protein kinase 6; mitogen-activated protein kinase 6; p55-MAPK; p97mapk; p97-MAPK; Prkm4; Prkm6; protein kinase, mitogen activated kinase 4; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6; si:ch211-235f12.1
Host Species Chicken
Immunogen Synthetic peptide taken within amino acid region 480-530 on human Mitogen-activated protein kinase 6.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50772, 5597, 58840
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.