Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK6/ERK3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982212
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2212 Supplier Bioss Supplier No. BS12405R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblasts upon treatment with serum or phorbol esters. Tissue specificity: Highest expression in the skeletal muscle, followed by the brain. Also found in heart, placenta, lung, liver, pancreas, kidney and skin fibroblasts.

Specifications

Antigen MAPK6/ERK3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAPK6
Gene Accession No. Q16659
Gene Alias 2610021I23Rik; BOS_10992; D130053K17Rik; DKFZp686F03189; Erk3; ERK-3; Erk-3 related; Extracellular signal-regulated kinase 3; extracellular signal-regulated kinase, p97; HsT17250; MAP kinase 6; MAP kinase isoform p97; MAPK 6; Mapk4; Mapk6; Mapk63; mitogen activated protein kinase 4; mitogen activated protein kinase 6; Mitogen-activated protein kinase 6; p55-MAPK; p97mapk; p97-MAPK; Prkm4; Prkm6; protein kinase, mitogen activated kinase 4; protein kinase, mitogen-activated 5; protein kinase, mitogen-activated 6; si:ch211-235f12.1
Gene Symbols MAPK6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAPK6/ERK3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5597
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.