Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK8/9/10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 280071AP150UL

Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This protein is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). Through its phosphorylation and nuclear localization, this kinase plays regulatory roles in the signaling pathways during neuronal apoptosis. Beta-arrestin 2, a receptor-regulated MAP kinase scaffold protein, is found to interact with, and stimulate the phosphorylation of this kinase by MAP kinase kinase 4 (MKK4). Cyclin-dependent kinase 5 can phosphorylate, and inhibit the activity of this kinase, which may be important in preventing neuronal apoptosis. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
MAPK8/9/10 | |
Polyclonal | |
Unconjugated | |
MAPK10 | |
c Jun N terminal kinase 3, FLJ12099, FLJ33785, JNK3, JNK3A, MAP kinase 10, MAP kinase p49 3F12, MAPK 10, MAPK10, MAPK8, MAPK9, p493F12, p54bSAPK, PRKM10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26414, 26419, 26420, 5599, 5601, 5602 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P45983, P45984, P53779, Q61831, Q91Y86, Q9WTU6 | |
MAPK10, MAPK8, MAPK9 | |
MAPK8/9/10 Fusion Protein Ag27823 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction