Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK8IP1 Monoclonal Antibody (OTI3A12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809896

Description
The protein encoded by this gene is a regulator of the pancreatic beta-cell function. It is highly similar to JIP-1, a mouse protein known to be a regulator of c-Jun amino-terminal kinase (Mapk8). This protein has been shown to prevent MAPK8 mediated activation of transcription factors, and decrease IL-1 beta and MAP kinase kinase 1 (MEKK1) induced apoptosis in pancreatic beta cells. This protein also functions as a DNA-binding transactivator of the glucose transporter GLUT2. RE1-silencing transcription factor (REST) is reported to repress the expression of this gene in insulin-secreting beta cells. This gene is found to be mutated in a type 2 diabetes family, and thus is thought to be a susceptibility gene for type 2 diabetes.Specifications
MAPK8IP1 | |
Monoclonal | |
Unconjugated | |
MAPK8IP1 | |
IB1, JIP-1, JIP1, PRKM8IP | |
Mouse | |
Affinity Chromatography | |
RUO | |
9479 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI3A12 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UQF2 | |
MAPK8IP1 | |
Full length human recombit protein of human MAPK8IP1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction