Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK9, Rabbit anti-Human, Clone: A11, Abnova™

Catalog No. 89335898 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-898 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-898 Supplier Abnova Corporation Supplier No. MAB19064
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen MAPK9
Applications Flow Cytometry
Classification Monoclonal
Clone A11
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA).
Gene mitogen-activated protein kinase 9
Gene Alias JNK-55/JNK2/JNK2A/JNK2ALPHA/JNK2B/JNK2BETA/PRKM9/SAPK/p54a/p54aSAPK
Gene Symbols MAPK9
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding T183/Y185 of human MAPK9/MAPK8.
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5601/5599
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.