Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPKAP Kinase 3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299449V
Description
MAPKAP Kinase 3 Polyclonal specifically detects MAPKAP Kinase 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MAPKAP Kinase 3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
3PKEC 2.7.11.1, Chromosome 3p kinase, EC 2.7.11, MAP kinase-activated protein kinase 3, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAP33pK, MAPKAPK-3, mitogen-activated protein kinase-activated protein kinase 3 | |
Produced in rabbits immunized with purified, recombinant MAPKAP Kinase 3 (NP_004626.1; Met1-Gln382). | |
0.1 mL | |
Protein Kinase | |
7867 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction