Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPKAP Kinase 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP29944950UL
Description
MAPKAP Kinase 3 Polyclonal antibody specifically detects MAPKAP Kinase 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MAPKAP Kinase 3 | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:500-1:2000 | |
3PKEC 2.7.11.1, Chromosome 3p kinase, EC 2.7.11, MAP kinase-activated protein kinase 3, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAP33pK, MAPKAPK-3, mitogen-activated protein kinase-activated protein kinase 3 | |
Produced in rabbits immunized with purified, recombinant Human MAPKAP Kinase 3 (Accession#: NP_004626.1; Met1-Gln382) | |
50 μg | |
Protein Kinase | |
7867 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction