Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPKAP Kinase 3 Antibody (SR1737), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322593100UL
Description
MAPKAP Kinase 3 Monoclonal antibody specifically detects MAPKAP Kinase 3 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
MAPKAP Kinase 3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
SR1737 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
3PKEC 2.7.11.1, Chromosome 3p kinase, EC 2.7.11, MAP kinase-activated protein kinase 3, MAPK-activated protein kinase 3, MAPKAP kinase 3, MAPKAP33pK, MAPKAPK-3, mitogen-activated protein kinase-activated protein kinase 3 | |
A synthesized peptide derived from human MAPKAP Kinase 3 (Uniprot #: Q16644) | |
100 μg | |
Protein Kinase | |
7867 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction