Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAPKAP1 Monoclonal Antibody (A6G10)
GREENER_CHOICE

Catalog No. PIMA544958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544958 Supplier Invitrogen™ Supplier No. MA544958
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAPKAP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Sin1 was initially identified as the human homolog of S. pombe SIN1. Recent evidence has shown that it identical to Mip1, a protein that interacts with MEKK2, a member of the mitogen-activated protein kinase (MAPK) intracellular signaling network. MAPKAP1 is thought to prevent MEKK2 activation and dimerization by forming a complex with the inactive and non-phosphorylated MEKK2, thereby blocking the JNK1/2, ERK1/2, p38 and ERK5 MAPKs. MAPKAP1 has also been shown to play a role in the TOR signaling process, a pathway that is involved in controlling cell growth and proliferation in response to environmental cues such as nutrients, growth factors and hormones. Experiments showed that MAPKAP1 helped to maintain the TOR/rictor assembly but not the TOR/RAPTOR complex, which allowed specific phosphorylation of Akt, a kinase that is believed to couple the growth factor-PI3K signaling pathway to the nutrient-regulated TOR signaling pathway. Multiple alternatively spliced isoforms of MAPKAP1 have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPKAP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A6G10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MAPKAP1
Gene Accession No. Q9BPZ7
Gene Alias AI591529; D230039K05Rik; im:7143088; JC310; MAPK associated protein 1; mapkap1; MEKK2-interacting protein 1; MGC2745; Mip1; MIP1SIN1SIN1b; mitogen activated protein kinase associated protein 1; mitogen-activated protein kinase 2-associated protein 1; mitogen-activated protein kinase associated protein 1; mSIN1; ras inhibitor MGC2745; RP11-269P11.1; SAPK interacting protein; SAPK-interacting protein 1; si:ch73-242f23.1; SIN1; SIN1b; SIN1g; stress-activated map kinase interacting protein 1; stress-activated map kinase-interacting protein 1; stress-activated protein kinase-interacting 1; target of rapamycin complex 2 subunit MAPKAP1; TORC2 subunit MAPKAP1
Gene Symbols MAPKAP1
Host Species Mouse
Immunogen Recombinant protein within human SIN1 aa 350-522.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79109
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.