Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPKAPK2 Antibody (7H4.2), Alexa Fluor™ 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229767AF488
Description
MAPKAPK2 Monoclonal antibody specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
MAPKAPK2 | |
Monoclonal | |
Alexa Fluor 488 | |
EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 | |
KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. | |
0.2 mL | |
Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
9261 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
7H4.2 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction