Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPKAPK2 Antibody (7H4.2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP229767AF700
Description
MAPKAPK2 Monoclonal antibody specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MAPKAPK2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 7H4.2 | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 | |
| KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. | |
| 0.2 mL | |
| Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| 9261 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction