Learn More
Promega MAPKAPK2 Kinase Enzyme System, 1mg
Catalog No. PRV6443 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6443 1 mg
1 options
Catalog No. PRV6443 Supplier Promega Supplier No. V6443
Add to Cart
Add to Cart
Request a Quote
Recombinant human MAPKAPK2 (46-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. MAPKAPK2 (MAPKAP kinase2) is a Ser/Thr protein kinase that is regulated via direct phosphorylation by p38 MAP kinase.
  • Easily Screen and Profile MAPKAPK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg