Learn More
Invitrogen™ MAPKAPK2 Recombinant Rabbit Monoclonal Antibody (4G7)

Description
MAPKAPK2 Recombinant Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | MAPKAPK2 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4G7 |
| Concentration | 0.73 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MAPKAPK2 |
| Gene Accession No. | P49137, P49138 |
| Gene Alias | AA960234; MAP kinase-activated protein kinase 2; MAPK activated protein kinase 2; MAPK-activated protein kinase 2; MAPKAP kinase 2; Mapkapk2; MAPKAP-K2; MAPKAPK-2; mitogen-activated protein kinase-activated protein kinase 2; MK2; MK-2; Rps6kc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.