Learn More
Promega MAPKAPK3 Kinase Enzyme System, 10microg
Catalog No. PRV4026 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4026 10 μg
1 options
Catalog No. PRV4026 Supplier Promega Supplier No. V4026
Recombinant full-length human MAPKAPK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MAPKAPK3 is closely related to MAPKAPK2 sharing 72% nucleotide and 75% amino acid identity.
  • Easily Screen and Profile MAPKAPK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C