Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPKAPK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598430 Supplier Thermo Scientific Supplier No. PA598430
Add to Cart
Add to Cart

MAPKAPK3 Polyclonal Antibody

MAPKAPK3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPKAPK3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAPKAPK3
Gene Accession No. Q16644
Gene Alias 3PK; AI874665; chromosome 3p kinase; MAP kinase-activated protein kinase 3; MAPK-activated protein kinase 3; MAPKAP kinase 3; MAPKAP3; MAPKAPK3; MAPKAP-K3; MAPKAPK-3; MapkKapk3; mitogen activated protein kinase-activated protein kinase 3; mitogen-activated protein kinase-activated protein kinase 3; MK3; MK-3; OTTHUMP00000210806; OTTHUMP00000210807
Gene Symbols MAPKAPK3
Host Species Rabbit
Immunogen Recombinant Human MAP kinase-activated protein kinase 3 protein (1-240AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7867
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.