Learn More
Promega MAPKAPK5 Kinase Enzyme System, 1mg
Catalog No. PRV6445 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6445 1 mg
1 options
Catalog No. PRV6445 Supplier Promega Supplier No. V6445
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human MAPKAPK5 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MAPKAPK5 is a member of the serine/threonine kinase family that responds to cellular stress and proinflammatory cytokines.
  • Easily Screen and Profile MAPKAPK5 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C