Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAPKBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109938
Description
Immunogen sequence: MAVEGSTITS RIKNLLRSPS IKLRRSKAGN RREDLSSKVT LEKVLGITVS GGRGLACDPR SGLVAYPAGC VVVLFNPRKH KQHHILNSSR KTITALAFSP.
MAP kinases play a significant role in many biological processes, including cell adhesion and spreading, cell differentiation and apoptosis. MAPKBP-1 (mitogen-activated protein kinase binding protein 1), also known as JNKBP-1, is a 1,514 amino acid protein that contains twelve WD repeats. Induced by TRAF2 (TNF receptor-associated factor 2) and Tak1 (TGF-beta-activated kinase 1), MAPKBP-1 is thought to act an adaptor protein for NFkappaB (nuclear factor kappa-B) activation. MAPKBP-1 interacts with JNK3 and may promote TRAF2 polyubiquitination. MAPKBP-1 exists as six alternatively spliced variants and is encoded by a gene located on human chromosome 15. Human chromosome 15 houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.
Specifications
| MAPKBP1 | |
| Polyclonal | |
| Unconjugated | |
| Mapkbp1 | |
| 2810483F24Rik; AW123212; JNK-binding protein 1; JNKBP1; JNKBP-1; Jun N-terminal kinase binding protein 1; Jun N-terminal kinase-binding protein 1; KIAA0596; Mapkbp1; mitogen activated protein kinase binding protein 1; mitogen-activated protein kinase binding protein 1; mitogen-activated protein kinase-binding protein 1; mKIAA0596 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23005, 26390 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60336, Q6NS57 | |
| Mapkbp1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MAPKBP1 (NP_001122080.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction