Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPRE1 Rabbit anti-Human, Polyclonal, Abnova™

Catalog No. 89035699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-699 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-699 Supplier Abnova Corporation Supplier No. PAB30206

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against synthetic peptide of human MAPRE1.

  • High specificity
  • High reactivity

Specifications

Antigen MAPRE1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of human MAPRE1.
Dilution Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4 (0.02% sodium azide).
Gene Accession No. Q15691
Gene Alias EB1/MGC117374/MGC129946
Gene Symbols MAPRE1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 130-180 of human MAPRE1.
Purification Method Immunoaffinity purification
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22919
Target Species Human
Content And Storage Store at -20°C on dry atmosphere, lyophilized antibodies are stable.
After reconstitution with deionized water, store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.