Learn More
MAPRE2 Mouse anti-Human, Clone: OTI1F3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502858

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.Specifications
MAPRE2 | |
Monoclonal | |
Unconjugated | |
MAPRE2 | |
CSCSC2, EB1, EB2, RP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10982 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1F3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15555 | |
MAPRE2 | |
Full length human recombit protein of human MAPRE2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.