Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAPRE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582383 Supplier Invitrogen™ Supplier No. PA582383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YDGKDYNPLL ARQGQDVAPP PNPGDQIFNK SKKLIGTAVP QRTSPTGPKN MQTSGRLSNV APPCILRKNP PSARNGGHET DAQILELNQQ LVDLKLTVDG LEKERDFYFS KLR Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

EB3 is a 282 amino acid homologue of EB1, a protein known to bind APC (adenomatous polyposis coli). It has been shown to bind to tubulin in vitro and microtubules in vivo. Northern blotting shows expressions to be mainly in brain, and to a lesser degree in muscle. EB3 interacts with APCL, a homologue of the tumor suppressor APC, and this interaction may be involved in the stabilization or extension of microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAPRE3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAPRE3
Gene Accession No. Q9UPY8
Gene Alias AI790651; APC binding protein; EB1 protein family member 3; EB2; EB3; EBF3; EBF3-S; End-binding protein 3; Mapre3; microtubule associated protein RP/EB family member 3; microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3
Gene Symbols MAPRE3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAPRE3. Recombinant protein control fragment (Product #RP-89599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.