Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPT (phospho T231) Rabbit anti-Human, Mouse, Rabbit, Polyclonal , Abnova™

Catalog No. 89956767 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-767 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-767 Supplier Abnova Corporation Supplier No. PAB29608
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAPT.

This gene encodes the microtubule-associated protein tau (MAPT) whose transcript undergoes complex, regulated alternative splicing, giving rise to several mRNA species. MAPT transcripts are differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type. MAPT gene mutations have been associated with several neurodegenerative disorders such as Alzheimer's disease, Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy. [provided by RefSeq]

Specifications

Antigen MAPT (phospho T231)
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of human MAPT.
Dilution Immunohistochemistry (1:50-1:100) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Alias DDPAC/FLJ31424/FTDP-17/MAPTL/MGC138549/MSTD/MTBT1/MTBT2/PPND/TAU
Gene Symbols MAPT
Host Species Rabbit
Immunogen A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T231 of human MAPT.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4137
Test Specificity MAPT (phospho T231) polyclonal antibody detects endogenous level of MAPT only when phosphorylated at threonine 231.
Target Species Human, Mouse, Rabbit
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.