Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Marapsin/Pancreasin Antibody (107), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290056PE
Description
Marapsin/Pancreasin Monoclonal antibody specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Marapsin/Pancreasin | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
83886 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
107 | |
Western Blot, Immunoprecipitation | |
CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27 | |
Rabbit immunized with purified, recombinant Human Marapsin/Pancreasin (Uniprot#: Q9BQR3, Met1-Lys290) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction