Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Marapsin/Pancreasin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32124925UL
Description
Marapsin/Pancreasin Polyclonal antibody specifically detects Marapsin/Pancreasin in Human samples. It is validated for ImmunofluorescenceSpecifications
Marapsin/Pancreasin | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPNMarapsin, Pancreasin, protease, serine 27, serine protease 27 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: NTSETSLYQVLLGARQLVQPGPHAMYARVRQVESNPLYQGTASSADVALVELEAPVPFTNYILPVCLPDPSVIFETGMNC | |
25 μg | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
83886 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction