Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Marburg Virus VP40 Monoclonal Antibody (3543)
GREENER_CHOICE

Catalog No. PIMA547504
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547504 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547504 Supplier Invitrogen™ Supplier No. MA547504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Specific for the VP40 antigen of the virus. Reactive with several native strains. Negative for EBOLA. Non-reactive with mammalian cells (ICC/IF). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Marburg virus (MBG) is a representative of Filoviridae family of RNA containing viruses. MBG is an exceptionally dangerous pathogen, which induces a severe contagious and highly lethal (53-88%) febrile disease with hemorrhagic syndrome. The name thread viruses' is based on their morphology. MBG made its appearance in 1967 in the form of frightening nosocomial outbreak, initially among polio vaccine production workers in Germany in contact with Ugandan green monkeys and their kidney tissues. The patho-and immuno-genesis of MBG fever have so far been little studied, no specific agents or treatment methods have been developed. VP40 is essential for assembly and release of newly copied viruses and also suppresses immune signaling in the infected cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Marburg Virus VP40
Applications ELISA
Classification Monoclonal
Clone 3543
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Immunogen Proprietary sequence.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C or -20°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.