Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCH1 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5144822
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144822 Supplier Thermo Scientific Supplier No. PA5144822

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: MLGWCEAIAR NPHRIPNNTR TPEISGDLAD ASQTSTLNEK SPGRSASRSS N Antigen sequence identity: Mouse 94%; Rat 31%

MARCH1 Polyclonal antibody specifically detects MARCH1 in Human samples. It is validated for Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARCH1
Gene Accession No. Q8TCQ1
Gene Alias 1-Mar, 2900024D24Rik, BB085186, E3 ubiquitin-protein ligase MARCH1, MARCH1, MARCHF1, MARCH-I, membrane associated ring finger 1, membrane associated ring-CH-type finger 1, membrane-associated ring finger (C3HC4) 1, membrane-associated ring finger (C3HC4) 1, E3 ubiquitin protein ligase, membrane-associated RING finger protein 1, Membrane-associated RING-CH protein I, RGD1305148, RING finger protein 171, RING-type E3 ubiquitin transferase MARCH1, RNF171
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1-51.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55016
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.