Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554763 Supplier Invitrogen™ Supplier No. PA554763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPDRESATEK DRGGSENAKK SPLSWDTKSE PRQEVGVNAE NVWSDCISVE HRPGTHDSEG YWKDYLNSSQ NSLDYFISGR PISPRSSVNS SYNPPAS Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 38%.

MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARCH10
Gene Accession No. Q8NA82
Gene Alias 10-Mar; 4933417C16Rik; AV039223; FLJ35757; March10; MARCH-X; membrane associated ring finger 10; membrane associated ring-CH-type finger 10; membrane-associated ring finger (C3HC4) 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; microtubule-associated E3 ubiquitin ligase; OTTMUSG00000002847; Probable E3 ubiquitin-protein ligase MARCH10; RGD1311692; RING finger protein 190; RING-type E3 ubiquitin transferase MARCH10; Rnf190; testis secretory sperm-binding protein Li 228n
Gene Symbols MARCH10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MARCH10. Recombinant protein control fragment (Product #RP-93424).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.