Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518853
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518853 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518853 Supplier Invitrogen™ Supplier No. PA518853
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MARCH10
Gene Accession No. Q5XIV2, Q8NA82
Gene Alias 10-Mar; 4933417C16Rik; AV039223; FLJ35757; March10; MARCH-X; membrane associated ring finger 10; membrane associated ring-CH-type finger 10; membrane-associated ring finger (C3HC4) 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; microtubule-associated E3 ubiquitin ligase; OTTMUSG00000002847; Probable E3 ubiquitin-protein ligase MARCH10; RGD1311692; RING finger protein 190; RING-type E3 ubiquitin transferase MARCH10; Rnf190; testis secretory sperm-binding protein Li 228n
Gene Symbols MARCH10
Host Species Goat
Immunogen Peptide with sequence C-SRNYPQPRTEE, from the internal region of the protein sequence (aa 778-788) (aa 778-788).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162333, 303596
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.