Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569239 Supplier Invitrogen™ Supplier No. PA569239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARCH11 Polyclonal Antibody for Western Blot

E3 ubiquitin-protein ligase that mediates polyubiquitination of CD4. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. May play a role in ubuquitin-dependent protein sorting in developmenting spermatids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MARCH11
Gene Accession No. A6NNE9
Gene Alias 11-Mar; 9630025C22; E3 ubiquitin-protein ligase MARCH11; MARCH11; MARCH-XI; membrane associated ring finger 11; membrane associated ring-CH-type finger 11; membrane-associated ring finger (C3HC4) 11; membrane-associated RING finger protein 11; Membrane-associated RING-CH protein XI; RGD1559945; RING-type E3 ubiquitin transferase MARCH11
Gene Symbols MARCH11
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human MARCH11.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441061
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.