Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569220 Supplier Invitrogen™ Supplier No. PA569220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

MARCH2 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases (EC 6.3.2.19). MARCH enzymes add ubiquitin (see MIM 191339) to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments. MARCH2 reduces surface accumulation of several glycoproteins and appears to regulate early endosome-to-trans-Golgi network (TGN) trafficking (Bartee et al., 2004 [PubMed 14722266]; Nakamura et al., 2005 [PubMed 15689499]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MARCHF2
Gene Accession No. Q99M02
Gene Alias 9530046H09Rik; E3 ubiquitin-protein ligase MARCH2; HSPC240; MARCH2; MARCHF2; MARCH-II; membrane associated ring finger 2; membrane associated RING-CH finger protein II; membrane associated ring-CH-type finger 2; membrane-associated ring finger (C3HC4) 2; membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase; membrane-associated RING finger protein 2; Membrane-associated RING-CH protein II; RGD1306395; RING finger protein 172; RING-type E3 ubiquitin transferase MARCH2; RNF172; syntaxin-6-binding protein
Gene Symbols MARCHF2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MARCH2 (aa 151-200).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224703
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.