Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCH3 Polyclonal Antibody
MARCH3 Polyclonal Antibody
Supplier: Thermo Scientific PA560351
Description
MARCH3 Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
MARCH3 | |
Polyclonal | |
Unconjugated | |
MARCH3 | |
6330411I15Rik; A530081L18Rik; AI843639; E3 ubiquitin-protein ligase MARCH3; MARCH3; MARCH-III; membrane associated ring finger 3; membrane associated ring-CH-type finger 3; membrane-associated ring finger (C3HC4) 3; membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase; membrane-associated RING finger protein 3; Membrane-associated RING-CH protein III; RING finger protein 173; RING-type E3 ubiquitin transferase MARCH3; RNF173 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UD3 | |
MARCH3 | |
Recombinant protein corresponding to Human MARCH3. Recombinant protein control fragment (Product #RP-99910). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction