Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116270
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116270 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116270 Supplier Invitrogen™ Supplier No. PA5116270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARCH3 Polyclonal Antibody for Western Blot

This gene encodes a member of the membrane-associated RING-CH (MARCH) family. The encoded protein is an E3 ubiquitin-protein ligase that may be involved in regulation of the endosomal transport pathway. [provided by RefSeq, Mar 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MARCH3
Gene Accession No. Q86UD3
Gene Alias 6330411I15Rik; A530081L18Rik; AI843639; E3 ubiquitin-protein ligase MARCH3; MARCH3; MARCH-III; membrane associated ring finger 3; membrane associated ring-CH-type finger 3; membrane-associated ring finger (C3HC4) 3; membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase; membrane-associated RING finger protein 3; Membrane-associated RING-CH protein III; RING finger protein 173; RING-type E3 ubiquitin transferase MARCH3; RNF173
Gene Symbols MARCH3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 224-251 amino acids from the C-terminal region of human MARH3.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115123
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.