Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARCH3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101570
Description
Antibody detects endogenous levels of total MARCH3.
This gene encodes a member of the membrane-associated RING-CH (MARCH) family. The encoded protein is an E3 ubiquitin-protein ligase that may be involved in regulation of the endosomal transport pathway. [provided by RefSeq, Mar 2013].
Specifications
MARCH3 | |
Polyclonal | |
Unconjugated | |
MARCH3 | |
6330411I15Rik; A530081L18Rik; AI843639; E3 ubiquitin-protein ligase MARCH3; MARCH3; MARCH-III; membrane associated ring finger 3; membrane associated ring-CH-type finger 3; membrane-associated ring finger (C3HC4) 3; membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase; membrane-associated RING finger protein 3; Membrane-associated RING-CH protein III; RING finger protein 173; RING-type E3 ubiquitin transferase MARCH3; RNF173 | |
Rabbit | |
Affinity chromatography | |
RUO | |
115123, 320253, 364878 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5XIE5, Q86UD3, Q8BRX9 | |
MARCH3 | |
A synthesized peptide derived from human MARCH3(Accession Q86UD3), corresponding to amino acid residues L115-L165. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction