Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARCH4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553149
Description
Immunogen sequence: WKVLNYDKTK DLEDQKAGGR TNPRTSSSTQ ANIPSSEEET AGTPAPEQGP AQAAGHPSGP LSHHHCAYTI LHILSHLRPH EQRSP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
RNF174 (MARCH4) is a multi-pass membrane protein mainly found in the membrane of Golgi apparatus. It is a complex enzyme with a RING-CH-type zinc finger. It acts as a ubiquitin-protein ligase mainly involved in third step of ubiquitin conjugation. The RINGv-type zinc finger domain is required for E3 ligase activity in which the E3 ubiquitin-protein ligase mediates ubiquitination of MHC-I and CD4, and promote their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. It is mainly expressed in brain and placenta.
Specifications
| MARCH4 | |
| Polyclonal | |
| Unconjugated | |
| MARCH4 | |
| BC056494; E3 ubiquitin-protein ligase MARCH4; Kiaa1399; MARCH4; MARCHF4; MARCH-IV; membrane associated ring finger 4; membrane associated ring-CH-type finger 4; membrane-associated ring finger (C3HC4) 4; membrane-associated ring finger (C3HC4) 4, E3 ubiquitin protein ligase; membrane-associated RING finger protein 4; Membrane-associated RING-CH protein IV; mKIAA1399; RING finger protein 174; RING-type E3 ubiquitin transferase MARCH4; RNF174 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57574 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2E8 | |
| MARCH4 | |
| Recombinant protein corresponding to Human MARCH4. Recombinant protein control fragment (Product #RP-90983). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction