Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116198
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116198 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116198 Supplier Invitrogen™ Supplier No. PA5116198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MARCH4 Polyclonal Antibody for Western Blot

RNF174 (MARCH4) is a multi-pass membrane protein mainly found in the membrane of Golgi apparatus. It is a complex enzyme with a RING-CH-type zinc finger. It acts as a ubiquitin-protein ligase mainly involved in third step of ubiquitin conjugation. The RINGv-type zinc finger domain is required for E3 ligase activity in which the E3 ubiquitin-protein ligase mediates ubiquitination of MHC-I and CD4, and promote their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. It is mainly expressed in brain and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH4
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MARCH4
Gene Accession No. Q9P2E8
Gene Alias BC056494; E3 ubiquitin-protein ligase MARCH4; Kiaa1399; MARCH4; MARCHF4; MARCH-IV; membrane associated ring finger 4; membrane associated ring-CH-type finger 4; membrane-associated ring finger (C3HC4) 4; membrane-associated ring finger (C3HC4) 4, E3 ubiquitin protein ligase; membrane-associated RING finger protein 4; Membrane-associated RING-CH protein IV; mKIAA1399; RING finger protein 174; RING-type E3 ubiquitin transferase MARCH4; RNF174
Gene Symbols MARCH4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 302-331 amino acids from the C-terminal region of human MARCH4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57574
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.