Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525584
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525584 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525584 Supplier Invitrogen™ Supplier No. PA525584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, chicken and Xenopus based on sequence homology.

MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MARCH5
Gene Accession No. Q3KNM2, Q9NX47
Gene Alias 1810015H18Rik; 2310008I22Rik; 2700055A20Rik; 5730499H23Rik; E130202O05Rik; E3 ubiquitin-protein ligase MARCH5; March5; Marchv; MARCH-V; membrane associated ring finger 5; membrane associated ring-CH-type finger 5; membrane-associated ring finger (C3HC4) 5; membrane-associated RING finger protein 5; Membrane-associated RING-CH protein V; mitochondrial ubiquitin ligase; MITOL; RING finger protein 153; RING-type E3 ubiquitin transferase MARCH5; RNF153
Gene Symbols MARCH5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 4-32 amino acids from the N-terminal region of human MARCH5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54708, 69104
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.