Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARCH5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566025
Description
Immunogen sequence: PDQALQQMLDR SCWVCFATDE DDRTAEWVRP CRCRGSTKWV HQACLQRWVD EKQRGNSTAR VACPQCNAEY LIVFPKLGPV VYVLDLA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2.
Specifications
MARCH5 | |
Polyclonal | |
Unconjugated | |
MARCH5 | |
1810015H18Rik; 2310008I22Rik; 2700055A20Rik; 5730499H23Rik; E130202O05Rik; E3 ubiquitin-protein ligase MARCH5; March5; Marchv; MARCH-V; membrane associated ring finger 5; membrane associated ring-CH-type finger 5; membrane-associated ring finger (C3HC4) 5; membrane-associated RING finger protein 5; Membrane-associated RING-CH protein V; mitochondrial ubiquitin ligase; MITOL; RING finger protein 153; RING-type E3 ubiquitin transferase MARCH5; RNF153 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54708 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NX47 | |
MARCH5 | |
Recombinant Human MARCH5. Recombinant protein control fragment (Product #RP-106715). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction