Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRMT61A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588013 Supplier Invitrogen™ Supplier No. PA588013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSFVAYEELI KEGDTAILSL GHGAMVAVRV QRGAQTQTRH GVLRHSVDLI GRPFGSKVTC GRGGWVYVLH PTPELWTLNL PHRTQILYST DIALITMMLE LRPGSVVCES GTGSGSVSHA IIRTIAPTGH LHTVEFHQQR AEKAREEFQE HRVGRWVTVR TQDVCRSGFG VSHVADAVFL DIPSPWEAVG HAWDALKVEG GRFCSFSPCI EQVQRTCQAL AARGFSELST LEVLPQVYNV RTVSLPPPDL GTGTDGPAGS DTSPFRSGTP MKEAVGHTGY LTFATKTPG; Positive Samples: Jurkat, BT-474, U-251MG, Mouse brain, Mouse pancreas, Mouse spleen, Mouse liver, Rat testis, Rat brain; Cellular Location: Nucleus.

Catalytic subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA (PubMed:16043508). Catalytic subunit of mRNA N(1)-methyltransferase complex, which mediates methylation of adenosine residues at the N(1) position of a small subset of mRNAs: N(1) methylation takes place in tRNA T-loop-like structures of mRNAs and is only present at low stoichiometries (PubMed:29107537, PubMed:29072297). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRMT61A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene TRMT61A
Gene Accession No. Q6AY46, Q80XC2, Q96FX7
Gene Alias 6720458F09Rik; AI606093; C14orf172; GCD14; GCD14 and PCMT domain containing protein RGD1359191; GCD14/PCMT domain containing protein RGD1359191; Gcd14p; hTRM61; mRNA methyladenosine-N(1)-methyltransferase catalytic subunit TRMT61A; RGD1359191; TRM61; Trmt61a; tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61A; tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRM61; tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61A; tRNA methyltransferase 61 homolog A; tRNA methyltransferase 61A; tRNA(m1A58)-methyltransferase subunit TRM61; tRNA(m1A58)-methyltransferase subunit TRMT61A; tRNA(m1A58)MTase subunit TRM61; tRNA(m1A58)MTase subunit TRMT61A
Gene Symbols TRMT61A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-289 of human TRMT61A (NP_6895202).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115708, 314462, 328162
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.