Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARCH9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen MARCH9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159692
SDP
View Documents
Novus Biologicals
NBP159692
100 μL
Each for $487.50
Only null left
Add to Cart
 
NBP15969220
SDP
View Documents
Novus Biologicals
NBP15969220UL
20 μL N/A N/A N/A
Description

Description

44264 Polyclonal specifically detects 44264 in Human samples. It is validated for Western Blot.
Specifications

Specifications

MARCH9
Polyclonal
Rabbit
Zinc Finger
E3 ubiquitin-protein ligase MARCH9, EC 6.3.2.-, FLJ36578, MARCH-IX, membrane-associated ring finger (C3HC4) 9, Membrane-associated RING finger protein 9, Membrane-associated RING-CH protein IX, RNF179RING finger protein 179
Synthetic peptides corresponding to MARCH9(membrane-associated ring finger (C3HC4) 9) The peptide sequence was selected from the middle region of 40246 (NP_612405). Peptide sequence SRTGPTSGATSRPPAAQRMRTLLPQRCGYTILHLLGQLRPPDARSSSHSG.
Primary
Western Blot
Unconjugated
RUO
Q86YJ5
92979
IgG
38 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.