Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MARCH9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588012 Supplier Invitrogen™ Supplier No. PA588012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTAGKSGPRN SRTGPTSGAT SRPPAAQRMR TLLPQRCGYT ILHLLGQLRP PDARSSSHSG REVVMRVTTV; Positive Samples: U-251MG, A-549, 293T, Mouse intestine, Mouse kidney, Mouse eye, Rat kidney, Rat eye; Cellular Location: Golgi apparatus membrane, Lysosome membrane, Multi-pass membrane protein.

MARCH9 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases (EC 6.3.2.19). MARCH enzymes add ubiquitin to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments. MARCH9 induces internalization of several membrane glycoproteins and directs them to the endosomal compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MARCH9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MARCH9
Gene Accession No. Q86YJ5
Gene Alias 9-Mar; AI608492; BC019560; E3 ubiquitin-protein ligase MARCH9; MARCH9; MARCH-IX; membrane associated ring finger 9; membrane associated ring-CH-type finger 9; membrane-associated ring finger (C3HC4) 9; membrane-associated ring finger 9; membrane-associated RING finger protein 9; Membrane-associated RING-CH protein IX; RING finger protein 179; RING-type E3 ubiquitin transferase MARCH9; RNF179
Gene Symbols MARCH9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 277-346 of human MARCH9 (NP_6124052).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92979
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.