Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCKS like protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MARCKS like protein |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MARCKS like protein Polyclonal specifically detects MARCKS like protein in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
MARCKS like protein | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
65108 | |
IgG | |
Affinity purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
F52, mac-MARCKS, MacMARCKS, Macrophage myristoylated alanine-rich C kinase substrate, MARCKS-like 1, MARCKS-like protein 1, MARCKS-related protein, MLP1, MLPMARCKS-like protein, MRPMACMARCKS | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human MARCKS like protein (NP_075385.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title