Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARCKS Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA595638
Description
MARCKS Polyclonal Antibody for Western Blot, ICC/IF, IHC
MARCKS is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis.
Specifications
MARCKS | |
Polyclonal | |
Unconjugated | |
MARCKS | |
80K protein; 80K-L; 80K-L protein; FLJ14368; FLJ90045; KINC; Macs; MARCKS; MARCS; myristoylated alanine rich protein kinase C substrate; myristoylated alanine-rich C-kinase substrate; Myristoylated alanine-rich protein kinase C substrate; myristoylated alanine-rich protein kinase C substrate (MARCKS, 80K-L); phosphomyristin; PKCSL; PRKCSL; protein kinase C substrate 80 kDa protein; protein kinase C substrate, 80 kDa protein, light chain | |
Chicken | |
IgY fraction | |
RUO | |
4082 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
~20 mg/mL | |
PBS with 10mM sodium azide | |
P29966 | |
MARCKS | |
Recombinant human MARCKS expressed in and purified from E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction