Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCKSL1 Polyclonal Antibody
MARCKSL1 Polyclonal Antibody
Supplier: Thermo Scientific PA556495
Description
MARCKSL1 Polyclonal Antibody for ICC/IF, IHC

Specifications
MARCKSL1 | |
Polyclonal | |
Unconjugated | |
Marcksl1 | |
AL022768; AW215397; AW536807; brain protein F52; D4Bc1; F52; MACMARCKS; Mac-MARCKS; Macrophage myristoylated alanine-rich C kinase substrate; Macs2; Macs3; MARCKS like 1; Marcksl1; MARCKS-like 1; MARCKS-like protein 1; MARCKS-related protein; MLP; MLP1; Mrp; myristoylated alanine-rich C kinase substrate | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65108 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49006 | |
Marcksl1 | |
Recombinant protein corresponding to Human MARCKSL1. Recombinant protein control fragment (Product #RP-102571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction