Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MARCKSL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577147
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MARCKS (Myristoylated alanine-rich protein kinase C substrate), also designated 80K or 80K-L, is a 332 amino acid protein that localizes to the plasma membrane and functions as a major cellular substrate for protein kinase C (PKC). MARCKSL1 (MARCKS-like protein 1), also known as MacMARCKS, MLP, MRP, or F52, is a 195 amino acid protein that, like MARCKS, is a major substrate for PKC. Expressed in a variety of tissues with highest levels found in testis and uterus, MARCKSL1 participates in the coordination of membrane-cytoskeletal signaling events, including secretion, migration, phagocytosis and cell adhesion. Additionally, MARCKSL1 functions as a regulator of Integrin activation and is thought to regulate Integrin-dependent signal transduction pathways, especially those involved in macrophage spreading.
Specifications
MARCKSL1 | |
Polyclonal | |
Unconjugated | |
Marcksl1 | |
AL022768; AW215397; AW536807; brain protein F52; D4Bc1; F52; MACMARCKS; Mac-MARCKS; Macrophage myristoylated alanine-rich C kinase substrate; Macs2; Macs3; MARCKS like 1; Marcksl1; MARCKS-like 1; MARCKS-like protein 1; MARCKS-related protein; MLP; MLP1; Mrp; myristoylated alanine-rich C kinase substrate | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17357, 65108, 81520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P28667, P49006, Q9EPH2 | |
Marcksl1 | |
Recombinant full length Human MARCKSL1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction