Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCO Antibody (2359A), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB29561C
Description
MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
MARCO | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
2359A | |
Flow Cytometry, CyTOF-ready | |
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
Mouse myeloma cell line, NS0-derived mouse MARCO protein, Gln70-Ser518, Accession # Q60754 | |
0.1 mL | |
Immunology, Innate Immunity | |
8685 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction