Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCO Antibody (579511), PE/Cy5.5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2956PECY55
Description
MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow CytometrySpecifications
MARCO | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
579511 | |
Flow Cytometry | |
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
Mouse myeloma cell line NS0-derived recombinant mouse MARCO, Gln70-Ser518, Accession # Q60754 | |
0.1 mL | |
Immunology, Innate Immunity | |
8685 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction