Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCO Mouse anti-Human, Clone: PLK1, Novus Biologicals™

Mouse Monoclonal Antibody
$292.00 - $633.00
Specifications
Antigen | MARCO |
---|---|
Clone | PLK1 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
Description
MARCO Monoclonal antibody specifically detects MARCO in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)Specifications
MARCO | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
Unconjugated | |
RUO | |
PBS | |
8685 | |
IgG3 κ | |
Protein A purified |
PLK1 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen | |
Monoclonal | |
Mouse | |
Human | |
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
The antibody was raised by immunising BALB/c mice with human alveolar macrophages (AMs) and subsequent creation of hybridomas. | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title