Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MARCO Mouse anti-Human, HRP, Clone: PLK1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311998H
Description
MARCO Monoclonal antibody specifically detects MARCO in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
MARCO | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
8685 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
PLK1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
The antibody was raised by immunising BALB/c mice with human alveolar macrophages (AMs) and subsequent creation of hybridomas. | |
0.1 mL | |
Primary | |
Human | |
IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction